N,N-Dimethylformamid ≥99,8% (durch GC), ChromAR® für die Flüssigchromatographie, für die UV-Spektrophotometrie, Macron Fine Chemicals™
5362-25
:
- Packung:Glass bottle
- VE:2,5 L
- Seq:5096
- Environmentally Preferable:
- For Laboratory,Research,or Manufacturing Use:≥99.8%
- ACS - Assay (HCON(CH₃)₂)(by GC) (corrected for water) (total isomers and ethylbenzene):≥99.8 %
- ACS - Color (APHA):≤15
- ACS - Residue after Evaporation:≤5 ppm
- ACS - Titrable Acid (meq/g):≤0.0005
- ACS - Titrable Base (meq/g):≤0.003
- ACS - Water (H₂O)(by Karl Fischer titrn):≤0.05 %
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 270 nm:≤1.00
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 275 nm:≤0.30
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 295 nm:≤0.10
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 310 nm:≤0.05
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 340 - 400 nm:≤0.01
: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
:
:73.09 g/mol
:13,03269
:153 °C (1013 hPa)
:–61 °C
:0.949 g/cm³ (20 °C)
:57.5 °C (closed cup)
:Raumtemperatur
:200-679-5
:2265
:3,III
:Filtered through a 0.2 µm filter.
:01-2119475605-32
:GHS02
:GHS08
:GHS07
:68-12-2
:HCON(CH₃)₂
:MFCD00003284