
4-Chloro-3-(trifluoromethyl)aniline
Catalog # ACTIAS20935G100
Supplier: ACTIVATE SCIENTIFIC
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:195.57 g/mol
Boiling Pt:130…132 °C (27 mmHg)
Melting Pt:36…37 °C
Storage Temperature:Ambient
EINECS:206-277-6
UN:2811
ADR:6.1,III
:GHS07
CAS Number:320-51-4
Formula:C₇H₅ClF₃N
MDL Number:MFCD00007846