Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Methoxycinnamic acid
3-Methoxycinnamic acid
3-Methoxycinnamic acid
3-Methoxycinnamic acid
ACTIAS24448G100
:
ACTIVATE SCIENTIFIC
:
3-Methoxycinnamic acid
ACTIAS24448G100
:
ACTIVATE SCIENTIFIC
:
Pack type:
Glass bottle for solids
Pk:
100 g
Seq:
3401
:
:
178.19 g/mol
:
228-049-5
:
GHS07
:
6099-04-3
:
C₁₀H₁₀O₃
:
MFCD00004386