Guanidinium hydrochloride for molecular biology
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride
CAS Number:50-01-1
:
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026
:95.53 g/mol
Merck Index:14,4562
Melting Pt:178…185 °C
Density:1.34
EINECS:200-002-3
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together