
EDTA (ethylenediamine tetraacetic acid) cell culture reagent
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:60-00-4
:
Formula:C₁₀H₁₆N₂O₈
MDL Number:MFCD00003541
:292.25 g/mol
Merck Index:14,3517
Boiling Pt:614 °C (1013 hPa)
Melting Pt:∼ 245 °C (dec.)
Density:0.86 g/cm³ (20 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:200-449-4
REACH:01-2119486399-18
:GHS07
Related Information
Frequently Bought Together