4-Aminobutyric acid ≥99%
Catalog # J61307.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
CAS Number:56-12-2
:
Formula:C₄H₉NO₂
MDL Number:MFCD00008226
:103.12 g/mol
Merck Index:14,430
Boiling Pt:248 °C (1013 hPa)
Melting Pt:∼ 195 °C (dec.)
Density:1.21 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-258-6