2-Chlorocyclopentanone
Glass bottle for solids, 5 g
Catalog # ACTIAS21390G5
Marketsource
Supplier: ACTIVATE SCIENTIFIC
2-Chlorocyclopentanone
Catalog # ACTIAS21390G5
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 694-28-0
Jump to:
Product Details & Documents
- CAS Number:694-28-0
- Formula:C₅H₇ClO
- MDL Number:MFCD00001410
- Molecular Weight:118.56 g/mol
- Boiling Pt:72 °C (1013 hPa)
- Density:1.2061 g/cm³ (0 °C)
- Storage Temperature:Refrigerator
- EINECS:211-769-9
- UN:2810
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



