Barium carbonate-[¹³C] (98% ¹³C )
Catalog # CAMBCLM-466-5
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
UN:1564
ADR:6.1,II
CAS Number:51956-33-3
Formula:CBaO₃