2-Chloro-6-fluoro-5-methylphenylboronic acid
Catalog # ACTIAS51156G1
Supplier: ACTIVATE SCIENTIFIC
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:188.3917 g/mol
CAS Number:352535-86-5
Formula:C₇H₇BClFO₂
MDL Number:MFCD03701535