6-Hydroxy-m-anisidine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:20734-76-3
:
Formula:C₇H₉NO₂
MDL Number:MFCD06616911
:139.16 g/mol
Melting Pt:135…137 °C
:GHS06
:GHS07
Storage Temperature:Ambient
Frequently Bought Together