Order Entry
Switzerland
ContactUsLinkComponent
1,3-Diphenylacetone-p-toluenesulphonylhydrazone ≥98%, Sigma-Aldrich®
1,3-Diphenylacetone-p-toluenesulphonylhydrazone ≥98%, Sigma-Aldrich®
Catalog # 230308-10G
CAS Number:  
undefined
1,3-Diphenylacetone-p-toluenesulphonylhydrazone ≥98%, Sigma-Aldrich®
Catalog # 230308-10G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance (Form):
    Conforms to Requirements
  • Infrared spectrum:
    Conforms to Structure

Specification Test Results

About this item

 :
 :378.49 g/mol
Melting Pt:187...188 °C (lit.)
Storage Temperature:Room Temperature
EINECS:243-344-9
CAS Number:19816-88-7
Formula:C₂₂H₂₂N₂O₂S
MDL Number:MFCD00009645