Order Entry
Switzerland
ContactUsLinkComponent
N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine for synthesis, Sigma-Aldrich®
N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Hill Formula:
    C₁₄H₃₂N₂O₄
  • Chemical formula:
    [CH₃CH(OH)CH₂]₂NCH₂CH₂N[CH₂CH(OH)CH₃]₂
  • Molar Mass:
    292.41 g/mol
  • Boiling point:
    240 °C (13 hPa)
  • Density:
    1.03 g/cm³ (20 °C)
  • pH value:
    10.4 (10 g/l, H₂O, 20 °C)
  • Refractive Index:
    1.478 (20 °C, 589 nm)
  • Solubility:
    (20 °C) soluble
  • LD 50 oral:
    LD50 Rat 2000 mg/kg
  • Storage class:
    10 - 13 Other liquids and solids
  • WGK:
    WGK 3 highly water endangering

Specification Test Results

Related Information

Frequently Bought Together