
N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:102-60-3
:
Formula:C₁₄H₃₂N₂O₄
MDL Number:MFCD00004534
:292.42 g/mol
Merck Index:14,3599
Boiling Pt:240 °C (10 mmHg)
Density:1.030
Flash Pt:>110 °C (230 °F)
EINECS:203-041-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together