trans-4-(Aminomethyl)cyclohexanecarboxylic acid
Catalog # MOLEM77717564
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:157.21 g/mol
EINECS:214-818-2
CAS Number:1197-18-8
Formula:C₈H₁₅NO₂
MDL Number:MFCD00001466