
trans-4-(Aminomethyl)cyclohexanecarboxylic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1197-18-8
:
Formula:C₈H₁₅NO₂
MDL Number:MFCD00001466
:157.21 g/mol
Merck Index:14,9569
Melting Pt:>300 °C
EINECS:214-818-2
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together