2,4,6-Trichloroquinazoline
Catalog # ACTIAS2566G1
Supplier: ACTIVATE SCIENTIFIC
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:233.4818 g/mol
CAS Number:20028-68-6
Formula:C₈H₃Cl₃N₂
MDL Number:MFCD08445622