4-Nitro-2,3-lutidine-N-oxide
Catalog # ACTIAS24165G100
Supplier: ACTIVATE SCIENTIFIC
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:168.15 g/mol
CAS Number:37699-43-7
Formula:C₇H₈N₂O₂
MDL Number:MFCD00065172