ContactUsLinkComponent
Universal absorbent mat pads, New Pig
Universal absorbent mat pads, New Pig
Catalog # NEWPMAT204PK
Supplier:  NEW PIG
Universal absorbent mat pads, New Pig
Catalog # NEWPMAT204PK
Supplier:  NEW PIG
Specifications
  • Description:
    Absorbent mat pads, gray
  • L×W:
    381×508 mm
  • Absorbency:
    Up to 3,78 kg/bag
  • Pk:
    200
Specifications
About this item
Lasts two times longer than ordinary mats for fewer change-outs, absorbs everything, works everywhere. Eight layers of 100% PP are thermally bonded to make these as strongest mat on the market, so these will not rip, tear or fray even when saturated. Mats take on the properties of absorbed liquids - always take proper precautions when handling saturated absorbents.
  • Lightweight and flame resistant
  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Highly absorbent, fine-fibre construction won't leave liquids or fiber residue behind
  • Absorbs most common industrial liquids - oils, water, solvents, coolants and more
  • Pads are ideal for catching drips and soaking up spills
Absorbed leaks and drips blend right in, dark grey colour hides grime so mat stays on the job longer. Mats take on the properties of absorbed liquids - always take proper precautions when handling saturated absorbents.