Safety rigger boots, pull-on, Portwest WORK Neptune FW 75
Nome del fornitore: Portwest
Certificati
Informazioni su questo articolo
Waterproof boot made from PVC/nitrile upper and PVC/nitrile sole.
- Fur lining
- Shock-absorbent heel
- Protective steel toecap (200 J)
- Puncture resistant steel midsole
- Side tabs for easy donning
For wet and cold conditions.
Colour: Black/tan
Certifications: EN ISO 20345:2011; S5, SRC.
Frequently Bought Together