SPE columns, PCB analysis, CHROMABOND® NAN
CHROMABOND® NAN is a special combination phase for SPE analysis of PCBs from sludge.
- Sodium sulfate for removal of trace water (N)
- SiOH/AgNO₃ phase for removal of sulfur, sulfur-containing and polar compounds (A)
Application: PCB extraction from sludge (hexane extract).
Frequently Bought Together