Solid phase extraction cartridges, HyperSep™ Aminopropyl
Supplier: Thermo Fisher Scientific
Certificates
About this item
A polar sorbent for both polar and anion exchange interactions.
- Weak anion exchanger
- Enhanced retention of strong anions, such as sulfonic acids
- Excellent retention of drugs, metabolites and structural isomers
Frequently Bought Together