Orders LinkContactUsLinkComponent
Ion exchangers
Ion exchangers
Supplier:  Merck
Certificates
Ion exchangers
10 Options Available Below
Product Details & Documents
In general, ion exchangers consist of a cross-linked polymer matrix with a uniform distribution of fixed ionic groups sited throughout the resin structure. These must be balanced by a similar number of ions of the opposite charge, (counter ions) to maintain electrical neutrality. Cation exchangers therefore, exchange and enrich only cations, anion exchangers only anions. In contrast adsorber resins have a non ionic, but depending on the structure, a somewhat polar character, and don't adsorb stoichiometrically both: anions, cations, as well as uncharged compounds.
Possible applications are:
Trace enrichment by using chelating ion exchangers
Determination of total salt content of solutions and water by H+ exchange
Removals of interfering cations or anions
Chromatographic separation
Digestion of insoluble compounds
Application as a catalyst
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • 1.15965.0500
    Mixed bed exchanger, Amberlite® IRN-150, H+/OH-, anion exchange capacity >0,6 mval/ml, cation exchange capacity >0,7 mval/ml
    Mixed bed exchanger, Amberlite® IRN-150, H+/OH-, anion exchange capacity >0,6 mval/ml, cation exchange capacity >0,7 mval/ml
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - SFr. 131.00
    SFr. 131.00
    1 * 500 mL
       SFr. 131.00
  • 1.15165.0500
    Mixed bed exchanger, Amberlite® MB-6113, H+/OH-, anion exchange capacity >0,6 mval/ml, cation exchange capacity >0,5 mval/ml
    Mixed bed exchanger, Amberlite® MB-6113, H+/OH-, anion exchange capacity >0,6 mval/ml, cation exchange capacity >0,5 mval/ml
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - SFr. 190.00
    SFr. 190.00
    1 * 500 g
       SFr. 190.00
  • 1.15131.0500
    Strong acid cation exchanger, Amberlite® IR-120, H+, exchange capacity >1,7 mval/ml
    Strong acid cation exchanger, Amberlite® IR-120, H+, exchange capacity >1,7 mval/ml
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - SFr. 77.50
    SFr. 77.50
    1 * 500 mL
       SFr. 77.50
  • 1.15131.5000
    Strong acid cation exchanger, Amberlite® IR-120, H+, exchange capacity >1,7 mval/ml
    Strong acid cation exchanger, Amberlite® IR-120, H+, exchange capacity >1,7 mval/ml
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - SFr. 518.00
    SFr. 518.00
    1 * 5 L
       SFr. 518.00
  • 1.15966.0500
    Strong acid cation exchanger, Amberlite® IR-120, Na+, exchange capacity >1,9 mval/ml
    Strong acid cation exchanger, Amberlite® IR-120, Na+, exchange capacity >1,9 mval/ml
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - SFr. 92.30
    SFr. 92.30
    1 * 500 mL
       SFr. 92.30
  • 1.15635.0500
    Strong acid cation exchanger, Amberlyst® 15, H+, exchange capacity >1,75 mval/ml
    Strong acid cation exchanger, Amberlyst® 15, H+, exchange capacity >1,75 mval/ml
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - SFr. 201.00
    SFr. 201.00
    1 * 500 mL
       SFr. 201.00
  • 1.12463.0500
    Strong basic anion exchanger, Amberlite® IRA-402, Cl-, exchange capacity >0,9 mval/ml (Cl-)
    Strong basic anion exchanger, Amberlite® IRA-402, Cl-, exchange capacity >0,9 mval/ml (Cl-)
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - SFr. 130.00
    SFr. 130.00
    1 * 500 mL
       SFr. 130.00
  • 1.05242.0250
    Strong basic anion exchanger, Dowex® 1-X8, Cl-, exchange capacity >1,2 mval/ml
    Strong basic anion exchanger, Dowex® 1-X8, Cl-, exchange capacity >1,2 mval/ml
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - SFr. 502.00
    SFr. 502.00
    1 * 250 mL
       SFr. 502.00
  • 1.04767.0500
    Strong basic anion exchanger, Ion exchanger III, OH-, exchange capacity >0,9 mval/ml
    Strong basic anion exchanger, Ion exchanger III, OH-, exchange capacity >0,9 mval/ml
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - SFr. 376.00
    SFr. 376.00
    1 * 500 g
       SFr. 376.00
  • 1.15959.0500
    Weak acid cation exchanger, Amberlite® IRA-67, OH-, exchange capacity >1,5 mval/ml
    Weak acid cation exchanger, Amberlite® IRA-67, OH-, exchange capacity >1,5 mval/ml
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - SFr. 389.00
    SFr. 389.00
    1 * 500 g
       SFr. 389.00