Orders LinkContactUsLinkComponent
Columns, MicroBore
Supplier:   VARIAN
Columns, MicroBore
Supplier:  VARIAN
Certificates
Columns, MicroBore
Columns, MicroBore
14 Options Available Below
Product Details & Documents
The microbore columns are ideal for use with detectors requiring low flow rates such as some mass
spectrometers and with capillary LC systems. Easily accommodates high temperatures for better selectivity and resolution between impurities and the target oligo.
  • Thermally and chemically stable
  • Exceptional resolution and selectivity
  • Contain durable and resilient polymer particles that deliver reproducible results over longer lifetimes
  • High purity levels
  • Gives a highly reproducible material that is free from silanols and heavy metal ions.
This consists of a range of pore sizes and particle sizes, all with identical chemistry and fundamental adsorptive characteristics. The particles are inherently hydrophobic, therefore no bonded phase, alkyl ligand is required for reversed-phase separations. Columns within the extensive product range are suitable for nano/capillary separations, including both bottom-up and top-down proteomics, analytical separations, and preparative purifications.
Product Family Options
Product Information
  • Column Format
    Phase
    Pore size
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • Microbore
    PLRP-S
    100 Å
    3 µm
    1,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Microbore
    PLRP-S
    300 Å
    3 µm
    1,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Microbore
    PLRP-S
    100 Å
    3 µm
    1,00 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Microbore
    PLRP-S
    300 Å
    3 µm
    1,00 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Microbore
    PLRP-S
    100 Å
    5 µm
    1,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Microbore
    PLRP-S
    1000 Å
    5 µm
    1,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Microbore
    PL-SCX
    1000 Å
    5 µm
    1,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Microbore
    PL-SCX
    1000 Å
    5 µm
    1,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Microbore
    PLRP-S
    300 Å
    5 µm
    1,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Microbore
    PLRP-S
    4000 Å
    5 µm
    1,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Microbore
    PL-SCX
    4000 Å
    5 µm
    1,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Microbore
    PL-SCX
    4000 Å
    5 µm
    1,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Microbore
    PLRP-S
    1000 Å
    8 µm
    1,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Microbore
    PLRP-S
    4000 Å
    8 µm
    1,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST