Orders LinkContactUsLinkComponent
Disposable earplugs, 304
Disposable earplugs, 304
Disposable earplugs, 304
Disposable earplugs, 304
Disposable earplugs, 304
These striped PVC-free foam plugs are available in two sizes to ensure a proper and comfortable fit.
  • PU foam expands gently in the ear canal, reduces tendency to back out of the ear canal
  • Smooth, soil resistant skin prevents dirt build-up, promotes proper hygiene
  • Tapered bullet-shaped design easier to roll-down and insert
Suitable for medium/high noise work situations that demand a high degree of comfort, frequent changes, or where hygiene presents a problem for re-use.
SNR: 33 dB.
Certifications: EN 352-2:2002
Product Family Options
  • Description
    Colour
    Size
  • Earplugs, packed in pairs
    White/yellow
    S
    1 * 100 PAIRE
    1 * 100 PAIRE - SFr. 78.00
    SFr. 78.00
    1 * 100 PAIRE
       SFr. 78.00
  • Earplugs, packed in pairs
    White/yellow
    L
    1 * 100 PAIRE
    1 * 100 PAIRE - SFr. 78.00
    SFr. 78.00
    1 * 100 PAIRE
       SFr. 78.00