ContactUsLinkComponent
Beakers with handle, SteriWare®
Beakers with handle, SteriWare®
Catalog # SAMP8049H-2000
Supplier:  Sampling Systems
Supplier #: 8049H-2000
undefined
Beakers with handle, SteriWare®
Catalog # SAMP8049H-2000
Supplier:  Sampling Systems
Supplier #: 8049H-2000
Log in for availability
Select an Option
129 mm
Select an Option
2000 ml
Select an Option
Disposable jug
Select an Option
152 mm
Select an Option
01
Select an Option
20
About this item
HDPE, translucent.
  • Individually bagged
  • BSE/TSE-free
  • Full batch traceability
  • Non graduated
  • Available as gamma irradiated sterile (SAL 10-6)
Disposable beakers have a wide base which make them very stable. The handle is rounded to make it comfortable to hold.
Frequently Bought Together