ContactUsLinkComponent
Window thermometer
Window thermometer
Catalog # 620-1181
undefined
Window thermometer
Catalog # 620-1181
Specifications
  • Length:
    200 mm
  • Environmentally Preferable:
  • Filling:
    Blue spirit
  • Pk:
    1
  • Range:
    −50…+50 °C
Specifications
About this item
Vienna type window thermometer with metal fittings.