Safety shoes, slip-on, Jalbio
Supplier: Jallatte
Certificates
About this item
Slip-on safety shoes made from Aseptane™ upper (resistant to fluid projection) and Softane™/Néotril™ sole (heat resistant up to 300 °C).
- Aseptane™ upper is water repellent, breathable, washable at 30°C
- Anatomically shaped Confort-Tech insole; anti-static and antibacterial, with shock absorber heel
- Lining made from anti-abrasion and breathable fibre
- Slip resistant Triftane™ sole helps to prevent fatigue
- Perforated steel toecap (200 J)
Suitable for food industries and wet working conditions.
Colour: White
Certifications: EN 345-1; S2, HRO
Frequently Bought Together