Order Entry
Switzerland
ContactUsLinkComponent
Inserts for screw neck vials, N 13
Inserts for screw neck vials, N 13
Catalog # MANA702972
Supplier:  MACHEREY-NAGEL
undefined
Inserts for screw neck vials, N 13
Catalog # MANA702972
Supplier:  MACHEREY-NAGEL

Specifications

  • Cap included:
    No
  • Ø×H:
    6×40 mm
  • Capacity:
    0,30 ml
  • Colour:
    Clear
  • Description:
    Inserts, conical (needs metal spring)
  • Material:
    Glass
  • Seq:
    03
  • Pk:
    100

Specifications

About this item