Industrial balances, Ranger® 3000 compact bench scale
Nome del fornitore: OHAUS
Certificati
Informazioni su questo articolo
Simple to operate, multipurpose, compact scale for producing precise results rapidly. Application modes include weighing, percent weighing, parts counting, accumulation and dynamic weighing.
- Large backlit LCD
- Portable with ABS lightweight plastic housing (net weight 4,2 kg) and rechargeable battery
- Simple keyboard, five-key operation
- Stabilisation within 1 second
- Weigh below capability
Ordering information: More connectivity options available, including optional Ethernet, 2nd RS232 or USB and optional 2nd platform. Approved versions are also available. Please see the website or contact your local Avantor sales office for more details.
Frequently Bought Together