Capillary columns, CP-Sil PONA CB
Nome del fornitore: VARIAN
Certificati
Informazioni su questo articolo
CP-Sil PONA CB delivers accurate analysis of paraffins, olefins, naphthalenes and aromatics in complex hydrocarbon mixtures.
- Guaranteed hydrocarbon analysis according to ASTM (DHA method)
- Inert to polar compounds for highly accurate data
- Excellent reproducibility increases productivity
Temperature limits: 250/275 °C
Similar phases: Petrocol DH, SPB-1, 007-1, Rtx-1, MXT-1
Frequently Bought Together