Cleanroom gloves, polyisoprene, TouchNTuff® 83-300 / 83-500
Supplier: Ansell
Certificates
About this item
These single-use thin polymer gloves provide superior tactility and flexible protection for both workers and products in diverse wet and dry environments, requiring contamination control.
- Sterile or non sterile, powder-free, anti-slip finish
- Hand-specific and comfortable to wear for a longer period of time
- Do not contain natural rubber latex: Safe for latex-sensitive (Type I) laboratory professionals
- SureFit Technology™: Beaded cuff with adhesive band prevents cuff from rolling down
- Inner coating technology enables ease of dry and wet hand donning
- Anti-slip finish helps prevent slippage
Ideal for people working with potentially hazardous chemicals as the gloves are resistant to chemical splashes. Also suitable for use in ISO Class 5 or higher cleanroom environments.
Length: 305 - 310 mm
Thickness: 0,21 ±0,04 mm (palm, single wall)
Colour: White
EN 421, EN 374-2, Cat. III; AQL 1.0
Certifications: EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS, EN 421; Cat. III; AQL 0,65
Packaging: Non sterile version (83-300): 20 right and left hand gloves packed in separate compartments in a polybag; 1 polybag per vacuum-sealed polybag; 2 polybags per vacuum-sealed master polybag, 5 master polybags per case liner; 200 pairs per carton.
Sterile version (83-500): One pair per poly wallet, sealed in a poly envelope; 10 pairs per sealed polybag; 5 sealed polybags per master bag; 4 master bags per case liner; 200 pairs per carton. All packaging is printed with cleanroom-compatible IPA resistant ink. The polybags have an easy-tear opening. The gloves are triple bagged for superior contamination control.
Sterile version (83-500): One pair per poly wallet, sealed in a poly envelope; 10 pairs per sealed polybag; 5 sealed polybags per master bag; 4 master bags per case liner; 200 pairs per carton. All packaging is printed with cleanroom-compatible IPA resistant ink. The polybags have an easy-tear opening. The gloves are triple bagged for superior contamination control.
Specifications
- Catalog No:
- 112-3182
- 112-2785
- 112-2783
- 112-2784
- 112-2782
- 112-2787
- 112-3180
- 112-2786
- 112-3181
- 112-2789
- 112-3178
- 112-2788
- 112-3179
- 112-3176
- 112-3177
- 112-3175
- Model:
- 83-300
- 83-500
- 83-500
- 83-500
- 83-500
- 83-500
- 83-300
- 83-500
- 83-300
- 83-500
- 83-300
- 83-500
- 83-300
- 83-300
- 83-300
- 83-300
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cleanroom class:
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- Finish:
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- EN norm:
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS, EN 421
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS, EN 421
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS, EN 421
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS, EN 421
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS, EN 421
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS, EN 421
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS, EN 421
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS, EN 421
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS, EN 421
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS, EN 421
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS, EN 421
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS, EN 421
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS, EN 421
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS, EN 421
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS, EN 421
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS, EN 421
Specifications
Frequently Bought Together