


Hygromycin B
Catalog # EHERC14260000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:527.5 g/mol
Storage Temperature:Refrigerator
EINECS:250-545-5
UN:3462
ADR:6.1,I
:GHS06
:GHS05
:GHS08
CAS Number:31282-04-9
Formula:C₂₀H₃₇N₃O₁₃
MDL Number:MFCD06795479