Order Entry
Switzerland
ContactUsLinkComponent
Mercury standard solution, 100 mg/L Hg in 5% nitric acid standard for ICP-MS
Mercury standard solution, 100 mg/L Hg in 5% nitric acid standard for ICP-MS
Catalog # ACSDM324.5NP.L1
Supplier:  CPACHEM
CAS Number:  
Mercury standard solution, 100 mg/L Hg in 5% nitric acid standard for ICP-MS
Catalog # ACSDM324.5NP.L1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Mercury
  • Symbol:
    Hg

Specifications

About this item