Cerium standard solution, 1,000 mg/L Ce in dil. nitric acid standard for ICP
Supplier: CPACHEM
Certificates
Certificates
About this item
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Silicone Rotational viscosity standards
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Conductivity standards, KCl, referenced at 25 °C
Supplier: REAGECON
Accessories for pH/°C meter, handheld
Supplier: Testo
VWR®, Sulphate 1000 mg/L in water standard for ion chromatography
Supplier: VWR Chemicals
Cupric citrate solution 0.1 M USP Test Solution (TS)
Supplier: VWR Chemicals
Redox buffer solutions
Supplier: Mettler - Toledo
Supelco® Organic Reference Standard, Chlorophyll a
Supplier: Merck
pH buffer solutions
Supplier: Mettler - Toledo
VWR®, Sulphur, Certified Reference Materials, in odourless kerosene
Supplier: VWR Chemicals
VWR® ARISTAR®, ICP Multi-Element Standard Solutions
Supplier: VWR Chemicals