Vanadium standard solution, 10000 mg/L V in 5% sulphuric acid standard for ICP
Supplier: CPACHEM
Certificates
About this item
Frequently Bought Together
Sodium tungstate dihydrate ≥99%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Supelco® PESTANAL® Organic Reference Standard, Hexazinone
Supplier: Merck
Bendamustine Related Compound, Reference Standard
Supplier: USP
Decitabine, Reference Standard
Supplier: USP
200g 1 * 200 g
Supplier: LGC Standards PROMOCHEM
VWR®, Cold Cranking Simulator (CCS) viscosity standards
Supplier: VWR Chemicals
100 µg/mL in Water 1 * 1 mL
Supplier: EHRENSTORFER
Accessories for ion selective meters, LAQUAtwin
Supplier: HORIBA
Barium oxide ≥99.5% (metals basis excluding Sr) (max. 0.05% Sr)
Supplier: Thermo Fisher Scientific
D-(-)-Fructose
Supplier: EHRENSTORFER
23-EPI-26-Deoxyactein, Reference Standard
Supplier: USP
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
Pesticide reference standard, 4-Chloroaniline 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Supplier: EHRENSTORFER