Chrysene-[D12] 200 µg/ml (98% D), CIL
Catalog # PROOCILDLM-261-1.2
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:1719-03-5
- Description:Chrysene-[D12] 200 µg/ml (98% D)
- Formula:C₁₈D₁₂
- Description:Chrysene-[D12]