Order Entry
Switzerland
ContactUsLinkComponent
Selenium standard solution, 10 mg/L Se in dil. nitric acid standard for ICP-MS
Selenium standard solution, 10 mg/L Se in dil. nitric acid standard for ICP-MS
Catalog # 89191.180
Supplier:  CPACHEM
CAS Number:  
Selenium standard solution, 10 mg/L Se in dil. nitric acid standard for ICP-MS
Catalog # 89191.180
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Selenium
  • Symbol:
    Se

Specifications

About this item