Order Entry
Switzerland
ContactUsLinkComponent
 
Phosphorus standard solution, 100 mg/L P in water standard for ICP-MS
Catalog # 89169.180
Supplier:  CPACHEM
CAS Number:  
Phosphorus standard solution, 100 mg/L P in water standard for ICP-MS
Catalog # 89169.180
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Phosphorus
  • Symbol:
    P

Specifications

About this item