Order Entry
Switzerland
ContactUsLinkComponent
 
Yttrium standard solution, 100 mg/L Y in dil. nitric acid standard for ICP-MS
Catalog # 89149.180
Supplier:  CPACHEM
CAS Number:  
Yttrium standard solution, 100 mg/L Y in dil. nitric acid standard for ICP-MS
Catalog # 89149.180
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Yttrium
  • Symbol:
    Y

Specifications

About this item