Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

3-Bromobenzamide

Marketsource
Supplier:   COMBI-BLOCKS
3-Bromobenzamide
Supplier:  COMBI-BLOCKS
Marketsource
Certificates
3-Bromobenzamide

2 Options Available Below

Product Details & Documents
  • CAS Number:22726-00-7
  • Formula:C₇H₆BrNO
  • MDL Number:MFCD00017127
  • Molecular Weight:200.04 g/mol
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g

    Specifications:
    Marketsource
    Marketsource