Order Entry
Switzerland
Orders LinkContactUsLinkComponent
D-(-)-Penicillamine

D-(-)-Penicillamine

Glass bottle for solids, 5 g
Catalog # BACE4032629.0005
Marketsource
Supplier:   BACHEM BIOCHEMICA
D-(-)-Penicillamine
Catalog # BACE4032629.0005
Supplier:  BACHEM BIOCHEMICA
Marketsource
CAS Number:  52-67-5
D-(-)-Penicillamine

Glass bottle for solids, 5 g

CAS Number:  52-67-5
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  H-D-Pen-OH, (S)-2-Amino-3-mercapto-3-methylbutanoic acid, Penicillamine
  • CAS Number:52-67-5
  • Formula:C₅H₁₁NO₂S
  • MDL Number:MFCD00064302
  • Molecular Weight:149.2 g/mol
  • EINECS:200-148-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.