

HBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat), Novabiochem® zur Synthese
Supplier: Novabiochem (Part of Merck)
Certificates
About this item
CAS Number:94790-37-1
:
Formula:C₁₁H₁₆F₆N₅OP
MDL Number:MFCD00075445
:379.2 g/mol
ADR:4.1,II
:M-Clarity™ Qualitätsstufe = MQ300
EINECS:423-020-5
UN:1325
:GHS02
:GHS07
Storage Temperature:Kühlschrank
Frequently Bought Together