Tetrafluoroborate standard solution, 1000 mg/L BF4- in Water standard for ion chromatography
Supplier: CPACHEM
Certificates
Tetrafluoroborate standard solution, 1000 mg/L BF4- in Water standard for ion chromatography
Supplier: CPACHEM
About this item
Frequently Bought Together
VOC-Mix 61 - 1 * 1 ml
Supplier: EHRENSTORFER
Internal Standards Mixture
Supplier: EHRENSTORFER
Deltamethrin certified 1 * 250 mg
Supplier: EHRENSTORFER
Supelco® Organic Reference Standard, 2,2,2-Trifluoroethanol
Supplier: Merck
Sulfamethoxazole, Reference Standard
Supplier: USP
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
ADAPALENE RELATED COMPOUND D 1 * 50 mg
Supplier: Merck
Total organic carbon (TOC) standards
Supplier: REAGECON
METALLOID/HYDRIDE ELEMENTS IN 20% HCL, TR. HF 1 * 100 mL
Supplier: LGC Standards PROMOCHEM
Dibutyl ether
Supplier: EHRENSTORFER
Eicosapentaenoic Acid, Reference Standards
Supplier: USP
Supelco® Organic Reference Standard, Bicyclohexyl
Supplier: Merck
AMMONIUM CAL SOLUTION 10MG/L 1 * 500 mL
Supplier: YSI ENVIRONMENTAL
N-FORMYLKYNURENINE 1 * 100 mg
Supplier: Merck
Paeoniflorin, Reference Standard
Supplier: USP
Sulphomolybdic reagent R2 Reag. Ph. Eur. 1086400
Supplier: VWR Chemicals