Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Ladle

Ladle

130 ml, 350 mm, 79 mm
Catalog # GERB3033
Supplier:   GERBER FUNKE
Ladle
Catalog # GERB3033
Supplier:  GERBER FUNKE
Ladle
Ladle

130 ml, 350 mm, 79 mm

Price
41.30 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents

Scoop with long handle, made from stainless steel.

Specifications
  • Ø:
    79 mm
  • Capacity:
    130 ml
  • Length:
    350 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.