Order Entry
Switzerland
ContactUsLinkComponent
Hydrometer for yogurt and chocolate milk
Hydrometer for yogurt and chocolate milk
Catalog # GERB6670
Supplier:  GERBER FUNKE
undefined
Hydrometer for yogurt and chocolate milk
Catalog # GERB6670
Supplier:  GERBER FUNKE

Specifications

  • Description:
    Hydrometer for yogurt and chocolate milk
  • Length:
    220 mm
  • Division:
    0,001 g/ml
  • Pk:
    1
  • Range:
    1,030 - 1,060 g/ml

Specifications

About this item

Hydrometer for yoghurt and chocolate milk, in the body reading at the top, with thermometer.