ContactUsLinkComponent
SPE columns, CHROMABOND® CN
SPE columns, CHROMABOND® CN
Supplier:  MACHEREY-NAGEL
Certificates
undefined
SPE columns, CHROMABOND® CN
Supplier:  MACHEREY-NAGEL
Specifications
  • Catalog No:
  • MANA730063
  • MANA730061
  • Carbon load:
  • 5,5%
  • 5,5%
  • Endcapped:
  • No
  • No
  • pH range:
  • 2 - 8
  • 2 - 8
  • Surface area:
  • 500 m²/g
  • 500 m²/g