Isolator gloves, CSM
Supplier: PIERCAN
Certificates
About this item
These gloves made of chlorosulphonated polyethylene provide excellent resistance to chemicals, sterilisation agents (hydrogen peroxide) and sterilisation processes (gamma and beta radiation).
- High resistance to ozone and UV
- Excellent flexibility and dexterity
- Excellent behavior to ionising radiations
Ideal for life science applications (pharmaceutical/medical).
Certifications: EN 388:2016 (3X11X), EN ISO 374-1 Type B (AMP), EN ISO 374-5 VIRUS, EN 421; Cat. III
Specifications
- Catalog No:
- PIECE10330Y8E44
- PIECE10330Y9E44
- PIECE10330Y6E44
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Cuff style:
- Beaded
- Beaded
- Beaded