ContactUsLinkComponent
Emergency blanket
Emergency blanket
Catalog # 118-0252
Supplier:  ORKLA CARE
undefined
Emergency blanket
Catalog # 118-0252
Supplier:  ORKLA CARE
Specifications
  • Description:
    Emergency blanket, disposable
  • L×W:
    1600×2100 mm
  • Pk:
    10
Specifications
About this item
This blanket, made of polyester film coated with aluminium on one side, is intended for use in a first aid situation. It helps prevent hypothermia, assists in prevention of post-accident shocks, and protects the injured person from wind and moisture.