PIG® high-visibility yellow mat pads and rolls
Supplier: NEW PIG
Certificates
About this item
Hi-viz spill absorbents alert workers to slip hazards while cleaning up leaks.
- High-visibility yellow colour is easier to see in poorly lit areas to minimise tripping hazards; alerts workers to potential dangers and work in progress
- Eight layers of 100% polypropylene are thermally bonded to make PIG Mat the strongest mat on the market; won't rip, tear or fray even when saturated
- Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
- Highly absorbent, fine-fibre construction won't leave liquids or fibre residue behind
- Absorbs and retains oils, coolants, solvents and water
- Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat whilst on the job
- Rolls are ideal for covering large areas and soaking up leaks and overspray
Easy-tear perforations let you take only what you need, so you use less mat and save money. Heavy-weight construction delivers greater durability and absorbency for demanding tasks and large-volume cleanups.
Frequently Bought Together