PIG® spill response bucket kit
Fournisseur: NEW PIG
Certificats
À propos de cet article
Just grab and go for fast response to spills up to 13,8 litres.
- Container is UN-rated for shipping waste (UN1H2/Y30/S)
- PIG socks stop spreading spills; PIG mat absorbs quickly
- Prepacked container provides easy access to contents
- Bold Spill Kit lettering for quick identification
- Temporary disposal bags with ties help make cleanup easier
Tamperproof seals help prevent pilfering of spill response supplies.
Attention: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.
Frequently Bought Together